Ethylmethacrylat 98+% stabilisiert

Lieferant: Thermo Scientific
Gefahr

Synonyme: Methacrylsäureethylester, Methylpropensäureethylester, 2-Methylpropensäureethylester

Stabilisierung: Stabilisiert mit 4-Methoxyphenol (MEHQ) 10 -120 ppm

A17518.AK A17518.AU
A17518.AKEA 26.4 EUR
A17518.AK A17518.AU
Ethylmethacrylat 98+% stabilisiert
Ethylmethacrylat

Ethyl methacrylate is used to prepare copolymers of EMA on waxy maize starch and hydroxypropylstarch, potential application of these copolymers are as excipients for compressed non-disintegrating matrix tablets. It is also used to study reactions of n-butyl acrylate and EMA with ozone in the gas phase.


Purity: 98+% CAS No.: 97-63-2 Molecular Formula: CH2=C(CH3)CO2CH2CH3 Formula Weight: 114.15 Storage Temperature: 2°C to 8°C Flash Point: 18°(64°F) Refractive Index: 1.4140 Odour: Pungent Melting Point: -60° Boiling Point: 118-119° Beilstein Reference: 471201 UN No.: 2277 MDL No.: MFCD00009161

Formel: C₆H₁₀O₂
Molecular Weight: 114,14 g/mol
Siedepunkt: 118…119 °C
Schmelzpunkt: –60 °C
Dichte: 0,917
Flammpunkt: 18 °C (64 °F)
Lagertemperatur: Kühlschrank
MDL: MFCD00009161
CAS-Nummer: 97-63-2
EINECS: 202-597-5
UN: 2277
ADR: 3,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR