Natriumethanolat 21% in Ethanol, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Ethanol Natriumsalz, Natriumethylat, Natriumethoxid

230553-100ML 230553-500ML
SIAL230553-100MLEA 37.6 EUR
SIAL230553-100ML SIAL230553-500ML
Natriumethanolat 21% in Ethanol, Sigma-Aldrich®
Natriumethanolat
Formel: C₂H₅NaO
Molecular Weight: 68,05 g/mol
MDL: MFCD00012417
CAS-Nummer: 141-52-6
EINECS: 205-487-5
UN: 3274
ADR: 3,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR