Osmium(III)chlorid, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
Osmiumtrichlorid
M-Clarity™ Qualitätsstufe = MQ100
398594-100MG
398594-500MG
SIAL398594-100MGEA
69.3
EUR
SIAL398594-100MG
SIAL398594-500MG
Osmium(III)chlorid, Sigma-Aldrich®
Osmium(III) chloride
Formel:
Cl₃Os Molecular Weight: 296,59 g/mol Schmelzpunkt: 500 °C Dichte: 5,3 g/cm³ (20 °C) |
MDL:
MFCD00011148 CAS-Nummer: 13444-93-4 EINECS: 236-587-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...