Osmium(III)chlorid Hydrat, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Osmiumtrichlorid Hydrat

M-Clarity™ Qualitätsstufe = MQ100

209066-2.5G 209066-500MG
SIAL209066-2.5GEA 823 EUR
SIAL209066-2.5G SIAL209066-500MG
Osmium(III)chlorid Hydrat, Sigma-Aldrich®
Osmium(III)chlorid Hydrat
Formel: Cl₃Os·H₂O
Molecular Weight: 314,6 g/mol
MDL: MFCD00149815
CAS-Nummer: 14996-60-2
EINECS: 236-587-7
UN: 3260
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR