Acrylamid-Bisacrylamid (29:1) 40% aqueous solution
Lieferant: MP Biomedicals
Gefahr
Synonyme: Acrylamid-Bis
11BIAC2903 11BIAC2902
ICNA11BIAC2903EA 558 EUR
ICNA11BIAC2903 ICNA11BIAC2902
Acrylamid-Bisacrylamid (29:1) 40% aqueous solution
Acrylamid-Bis (32:1)
Lagertemperatur: Raumtemperatur
MDL: MFCD00080848
UN: 3426
ADR: 6.1 III