trans-2-Aminocyclohexanol Hydrochlorid 99%

Lieferant: Thermo Scientific
Achtung

206980050 206980250
ACRO206980050EA 60.7 EUR
ACRO206980050 ACRO206980250
trans-2-Aminocyclohexanol Hydrochlorid 99%
trans-2-Aminocyclohexanol Hydrochlorid
Formel: C₆H₁₄ClNO
Molecular Weight: 151,64 g/mol
Schmelzpunkt: 173…177 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00001490
CAS-Nummer: 5456-63-3

Bestellen

Spezifikation Testergebnisse

Appearance White to beige to light brown Crystalline powder
Infrared spectrum Conforms
Melting point 173 to 177 °C
Titration with HClO4 ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR