trans-2-Aminocyclohexanol Hydrochlorid, Sigma-Aldrich®
Lieferant: Merck
M-Clarity™ Qualitätsstufe = MQ100
222577-25G
222577-5G
SIAL222577-25GEA
227
EUR
SIAL222577-25G
SIAL222577-5G
trans-2-Aminocyclohexanol Hydrochlorid, Sigma-Aldrich®
trans-2-Aminocyclohexanol Hydrochlorid
Formel:
C₆H₁₄ClNO Molecular Weight: 151.64 g/mol Schmelzpunkt: 174…174 °C |
MDL:
MFCD00001490 CAS-Nummer: 5456-63-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...