trans-4-Aminocyclohexanol ≥98%

Lieferant: Thermo Fisher Scientific
Gefahr

B22365.06 B22365.14 B22365.22
B22365.06EA 34.1 EUR
B22365.06 B22365.14 B22365.22
trans-4-Aminocyclohexanol ≥98%
trans-4-Aminocyclohexanol

Purity: 98+% CAS No.: 27489-62-9 Molecular Formula: C6H13NO Formula Weight: 115.18 Melting Point: 110-113° UN No.: 3259 MDL No.: MFCD00067698

Formel: C₆H₁₃NO
Molecular Weight: 115.18 g/mol
Siedepunkt: 127 °C (14 mmHg)
Schmelzpunkt: 110…113 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00067698
CAS-Nummer: 27489-62-9
EINECS: 248-492-8
ADR: 8 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR