trans-4-Aminocyclohexanol Hydrochlorid, Sigma-Aldrich®
Lieferant: Merck
M-Clarity™ Qualitätsstufe = MQ100
263761-100G
263761-25G
SIAL263761-100GEA
187
EUR
SIAL263761-100G
SIAL263761-25G
trans-4-Aminocyclohexanol Hydrochlorid, Sigma-Aldrich®
trans-4-Aminocyclohexanol Hydrochlorid
Formel:
C₆H₁₄ClNO Molecular Weight: 151,64 g/mol |
MDL:
MFCD00012566 CAS-Nummer: 50910-54-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...