Triethylammoniumbromid 98%

Lieferant: Thermo Scientific
Achtung

Synonyme: Triethylamin Hydrobromid

A17926.22 A17926.36
A17926.22EA 39.4 EUR
A17926.22 A17926.36
Triethylammoniumbromid 98%
Triethylammonium bromide

Triethylamine hydrobromide is used to prepare dimethyl -alfa-diethylaminomaleate by reacting with dimethyl acetylenedicarbosylate. It is involved in the preparation of triphenylpropargylphosphonium bromide.


Purity: 98% CAS No.: 636-70-4 Molecular Formula: C16H15N.HBr Formula Weight: 182.10 Odour: Amine-like Melting Point: ca 250° dec. MDL No.: MFCD00054287

Formel: C₆H₁₅N·HBr
Molecular Weight: 182,1 g/mol
Schmelzpunkt: ∼ 250 °C (Zers.)
Lagertemperatur: Raumtemperatur
MDL: MFCD00054287
CAS-Nummer: 636-70-4
EINECS: 211-263-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR