Triethylammoniumchlorid 98%

Lieferant: Thermo Scientific
Achtung

Synonyme: Triethylamin Hydrochlorid

A17318.0B A17318.30
A17318.0BEA 98.1 EUR
A17318.0B A17318.30
Triethylammoniumchlorid 98%
Triethylammonium chloride

Purity: 98% CAS No.: 554-68-7 Molecular Formula: C6H15N.HCl Formula Weight: 137.65 Odour: Amine-like pH: 5 Melting Point: ca 260° dec. Beilstein Reference: 3906409 MDL No.: MFCD00012500

Formel: C₆H₁₆ClN
Molecular Weight: 137,65 g/mol
Schmelzpunkt: ∼ 260 °C (Zersetzung)
Dichte: 1,07
Lagertemperatur: Raumtemperatur
MDL: MFCD00012500
CAS-Nummer: 554-68-7
EINECS: 209-067-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR