Kaliumtrifluoracetat

Lieferant: FLUOROCHEM

Synonyme: Trifluoressigsäure Kaliumsalz

001216-100G 001216-25G
FLUO001216-100GEA 62 EUR
FLUO001216-100G FLUO001216-25G
Kaliumtrifluoracetat
Kaliumtrifluoracetat
Formel: C₂F₃KO₂
Molecular Weight: 152.11 g/mol
MDL: MFCD00013215
CAS-Nummer: 2923-16-2
EINECS: 220-877-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR