3-Chlor-4-hydroxybenzaldehyd
Lieferant: Thermo Scientific
RH00193.1
RH00193.10
RH00193.50
MAYBRH00193.1EA
0
EUR
MAYBRH00193.1
MAYBRH00193.10
MAYBRH00193.50
3-Chlor-4-hydroxybenzaldehyd
3-Chloro-4-hydroxybenzaldehyde
Formel:
C₇H₅ClO₂ Molecular Weight: 156,57 g/mol Siedepunkt: 251 °C (1013 hPa) Schmelzpunkt: 133…134 °C Dichte: 1,0404 g/cm³ (20 °C) |
MDL:
MFCD00016981 CAS-Nummer: 2420-16-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...