4-Hydroxybutanon 95%

Lieferant: Thermo Scientific
Achtung

Synonyme: 4-Hydroxy-2-butanone, 4-Hydroxybutan-2-one

L11456.14 L11456.22
L11456.14EA 51.4 EUR
L11456.14 L11456.22
4-Hydroxybutanon 95%
4-Hydroxybutanon

Purity: 95% CAS No.: 590-90-9 Molecular Formula: C4H8O2 Formula Weight: 88.11 Flash Point: 89°(192°F) Refractive Index: 1.4310 Boiling Point: 73-76°/12mm Beilstein Reference: 1737666 MDL No.: MFCD00059005

Formel: C₄H₈O₂
Molecular Weight: 88,11 g/mol
Siedepunkt: 73…76 °C (12 mmHg)
Dichte: 1,020
Flammpunkt: 89 °C (192 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00059005
CAS-Nummer: 590-90-9
EINECS: 209-693-6
UN: 1993
ADR: 3,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR