2,3-Pentandion ≥97%
Lieferant: Thermo Scientific
Synonyme:
Pentane-2,3-dione
A13116.14
A13116.22
A13116.14EA
34.2
EUR
A13116.14
A13116.22
2,3-Pentandion ≥97%
2,3-Pentandion
Purity: 97%
CAS No.: 600-14-6
Molecular Formula: C5H8O2
Formula Weight: 100.12
Storage Temperature: 2°C to 8°C
Flash Point: 18°(64°F)
Refractive Index: 1.4050
Odour: Odourless
Melting Point: -52°
Boiling Point: 110-112°
Beilstein Reference: 1699638
UN No.: 1224
MDL No.: MFCD00009313
Formel:
C₅H₈O₂ Molecular Weight: 100,12 g/mol Siedepunkt: 110 °C (1013 hPa) Schmelzpunkt: –52 °C Dichte: 0,96 g/cm³ (20 °C) Flammpunkt: 18 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD00009313 CAS-Nummer: 600-14-6 EINECS: 209-984-8 UN: 1224 ADR: 3,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...