3,4-Hexandion 94%
Lieferant: Thermo Scientific
|
Achtung
|
L06655.14
L06655.22
L06655.14EA
38.4
EUR
L06655.14
L06655.22
3,4-Hexandion 94%
3,4-Hexandion
Purity: 94%
CAS No.: 4437-51-8
Molecular Formula: C6H10O2
Formula Weight: 114.15
Flash Point: 29°(84°F)
Refractive Index: 1.4120
Melting Point: -10°
Boiling Point: 129-131°
Beilstein Reference: 1700837
UN No.: 1224
MDL No.: MFCD00010237
Formel:
C₆H₁₀O₂ Molecular Weight: 114,14 g/mol Siedepunkt: 129…131 °C Schmelzpunkt: –10 °C Dichte: 0,941 Flammpunkt: 29 °C (84 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00010237 CAS-Nummer: 4437-51-8 EINECS: 224-651-7 UN: 1224 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...