Ethylacetat ≥99,5% ACS

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: Essigester, Essigsäureethylester, EtOAc

31344.AP 31344.K2 31344.M1 31344.M4
31344.APEA 36.3 EUR
31344.AP 31344.K2 31344.M1 31344.M4
Ethylacetat ≥99,5% ACS
Ethylacetat

Purity: 99.5+% CAS No.: 141-78-6 Molecular Formula: C4H8O2 Formula Weight: 88.11 Flash Point: -3°(26°F) Physical Form: Liquid Density: 0.902 Refractive Index: 1.3720 Appearance: Colourless Odour: Sweet Melting Point: -84° Boiling Point: 77° Beilstein Reference: 506104 Merck Reference: 14,3757 UN No.: 1173 MDL No.: MFCD00009171

Formel: CH₃COOC₂H₅
Siedepunkt: 77 °C
Schmelzpunkt: –84 °C
Dichte: 0,902
Flammpunkt: –3 °C (26 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00009171
CAS-Nummer: 141-78-6
EINECS: 205-500-4
UN: 1173
ADR: 3,II
REACH: 01-2119475103-46
Merck Index: 14,3757
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=226aecffe42ffab125597552eff0ef7f88ef9983&vid=f5707ce0faee09ca10700419252c5841e0fbc01a&prid=web&clid=SAPDAM

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR