1,1,2,2-Tetrachlorethan ≥98%
Lieferant: Thermo Scientific
|
Gefahr
|
L04489.14
L04489.22
L04489.36
L04489.14EA
26.3
EUR
L04489.14
L04489.22
L04489.36
1,1,2,2-Tetrachlorethan ≥98%
1,1,2,2-Tetrachloroethane
Purity: 98+%
CAS No.: 79-34-5
Molecular Formula: C2H2Cl4
Formula Weight: 167.85
Refractive Index: 1.4935
Melting Point: -43°
Boiling Point: 142-146°
Beilstein Reference: 969206
Merck Reference: 14,9189
UN No.: 1702
MDL No.: MFCD00000848
Formel:
Cl₂HCCHCl₂ Molecular Weight: 167,85 g/mol Siedepunkt: 142…146 °C Schmelzpunkt: –43 °C Dichte: 1,596 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00000848 CAS-Nummer: 79-34-5 EINECS: 201-197-8 UN: 1702 ADR: 6.1,II Merck Index: 14,9189 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...