1,3-Dichloraceton 96%

Lieferant: Thermo Scientific
Gefahr

A14149.14 A14149.22 A14149.36
A14149.14EA 37.4 EUR
A14149.14 A14149.22 A14149.36
1,3-Dichloraceton 96%
1,3-Dichloraceton

Purity: 96% CAS No.: 534-07-6 Molecular Formula: C3H4Cl2O Formula Weight: 126.97 Storage Temperature: 2°C to 8°C Flash Point: 95°(203°F) Refractive Index: 1.4400 Odour: Pungent pH: 4.2 Melting Point: 40-44° Boiling Point: 172-173° Beilstein Reference: 605456 Merck Reference: 14,3052 UN No.: 2649 MDL No.: MFCD00000937

Formel: ClH₂CC(O)CH₂Cl
Molecular Weight: 126,97 g/mol
Siedepunkt: 172…173 °C
Schmelzpunkt: 40…44 °C
Dichte: 1,383
Flammpunkt: 95 °C (203 °F)
Lagertemperatur: Kühlschrank
MDL: MFCD00000937
CAS-Nummer: 534-07-6
EINECS: 208-585-6
UN: 2649
ADR: 6.1,II
Merck Index: 14,3052

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR