1-Chlor-1,1,2-trifluor-2-iodethan

Lieferant: FLUOROCHEM

Synonyme: 1-Chloro-2-iodo-1,1,2-trifluoroethane

004488-5G 004488-25G
FLUO004488-5GEA 104 EUR
FLUO004488-5G FLUO004488-25G
1-Chlor-1,1,2-trifluor-2-iodethan
1-Chlor-1,1,2-trifluor-2-iodethan
Formel: C₂HClF₃I
Molecular Weight: 244,38 g/mol
MDL: MFCD00042138
CAS-Nummer: 354-26-7
EINECS: 206-553-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR