5-Chlor-1-pentanol
Lieferant: FLUOROCHEM
|
Achtung
|
Synonyme:
5-Chloropentan-1-ol
008844-1G
008844-5G
FLUO008844-1GEA
31.1
EUR
FLUO008844-1G
FLUO008844-5G
5-Chlor-1-pentanol
5-Chlor-1-pentanol
Formel:
C₅H₁₁ClO Molecular Weight: 122.59 g/mol Siedepunkt: 52…53.5 °C (0.6 mmHg) Dichte: 1.046 g/cm³ (17 °C) |
MDL:
MFCD00039556 CAS-Nummer: 5259-98-3 EINECS: 226-067-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...