1,1,1,5,5,5-Hexafluoracetylaceton ≥98%
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
1,1,1,5,5,5-Hexafluor-2,4-pentandion
L01693.06
L01693.22
L01693.06EA
56.4
EUR
L01693.06
L01693.22
1,1,1,5,5,5-Hexafluoracetylaceton ≥98%
1,1,1,5,5,5-Hexafluoracetylaceton
Purity: 98+%
CAS No.: 1522-22-1
Molecular Formula: C5H2F6O2
Formula Weight: 208.06
Flash Point: 32°(89°F)
Refractive Index: 1.3340
Boiling Point: 69-71°
Beilstein Reference: 1790138
UN No.: 2920
MDL No.: MFCD00000426
Formel:
C₅H₂F₆O₂ Molecular Weight: 208,06 g/mol Siedepunkt: 69…71 °C Dichte: 1,486 Flammpunkt: 32 °C (89 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00000426 CAS-Nummer: 1522-22-1 EINECS: 216-191-0 UN: 2920 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...