Thallium(III)trifluoracetat ≥95%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Synonyme:
Trifluoressigsäure Thallium(III)salz
87913.04
87913.09
87913.18
87913.04EA
42.4
EUR
87913.04
87913.09
87913.18
Thallium(III)trifluoracetat ≥95%
Thallium(III)trifluoracetat
Purity: 95%
CAS No.: 23586-53-0
Molecular Formula: Tl(OOCCF3)3
Formula Weight: 543.42
Physical Form: Solid
Sensitivity: Hygroscopic
UN No.: 1707
MDL No.: MFCD00000414
Formel:
C₆F₉O₆Tl Molecular Weight: 543.43 g/mol Siedepunkt: 160 °C (1.5 mmHg) Schmelzpunkt: 213 °C (Zers.) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00000414 CAS-Nummer: 23586-53-0 EINECS: 245-761-1 UN: 1707 ADR: 6.1 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...