1-Iodperfluordecan ≥97%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
B21845.06
B21845.14
B21845.06EA
60.7
EUR
B21845.06
B21845.14
1-Iodperfluordecan ≥97%
1-Iodperfluordecan
Purity: 97%
CAS No.: 423-62-1
Molecular Formula: CF3(CF2)9I
Formula Weight: 645.98
Flash Point: None
Melting Point: 63-67°
Boiling Point: 195-200°
Beilstein Reference: 1810506
MDL No.: MFCD00001065
Formel:
C₁₀F₂₁I Siedepunkt: 195…200 °C Schmelzpunkt: 63…67 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00001065 CAS-Nummer: 423-62-1 EINECS: 207-030-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...