1H-1,2,4-Triazol-3-thiol
Lieferant: Thermo Scientific
Synonyme:
3-Mercapto-1H-1,2,4-triazol, 3-Mercapto-1,2,4-triazol
SB00858.1
SB00858.10
SB00858.50
SB00858-1
MAYBSB00858.1EA
0
EUR
MAYBSB00858.1
MAYBSB00858.10
MAYBSB00858.50
MAYBSB00858-1
1H-1,2,4-Triazol-3-thiol
1H-1,2,4-Triazol-3-thiol
Formel:
C₂H₃N₃S Molecular Weight: 101,13 g/mol Schmelzpunkt: 217…220 °C |
MDL:
MFCD00005229 CAS-Nummer: 3179-31-5 EINECS: 221-656-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...