Trihexylamin 97%

Lieferant: Thermo Scientific
Achtung

Synonyme: Tri-n-hexylamine, Tri(hex-1-yl)amine

B23716.18 B23716.30
B23716.18EA 43.2 EUR
B23716.18 B23716.30
Trihexylamin 97%
Trihexylamin

Purity: 97% CAS No.: 102-86-3 Molecular Formula: [CH3(CH2)5]3N Formula Weight: 269.52 Flash Point: 144°(291°F) Refractive Index: 1.4415 Melting Point: -75° Boiling Point: 263-265° Beilstein Reference: 1755828 UN No.: 3082 MDL No.: MFCD00009523

Formel: C₁₈H₃₉N
Molecular Weight: 269,51 g/mol
Siedepunkt: 263…265 °C
Schmelzpunkt: < –75 °C
Dichte: 0,798
Flammpunkt: 144 °C (291 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00009523
CAS-Nummer: 102-86-3
EINECS: 203-062-9
UN: 3082
ADR: 9,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR