Acetondimethylacetal ≥98%
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
2,2-Dimethoxypropan
A13810.0F
A13810.AE
A13810.AP
AB116057.0100
A13810.PI
A13810.0FEA
207
EUR
A13810.0F
A13810.AE
A13810.AP
ABCRAB116057.0100
A13810.PI
Acetondimethylacetal ≥98%
Acetondimethylacetal
Purity: 98%
CAS No.: 77-76-9
Molecular Formula: (CH3O)2C(CH3)2
Formula Weight: 104.15
Flash Point: -11°(12°F)
Physical Form: Liquid
Density: 0.848
Refractive Index: 1.3780
Appearance: Colourless
Odour: Sweet
Melting Point: -47°
Boiling Point: 79-81°
Beilstein Reference: 635678
UN No.: 3271
MDL No.: MFCD00008479
Formel:
C₅H₁₂O₂ Molecular Weight: 104,15 g/mol Siedepunkt: 79…81 °C Schmelzpunkt: –47 °C Dichte: 0,848 Flammpunkt: –11 °C (–12 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00008479 CAS-Nummer: 77-76-9 EINECS: 201-056-0 UN: 3271 ADR: 3,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...