1-Decanol ≥98%

Lieferant: Thermo Scientific
Achtung

Synonyme: 1-Decylalkohol, Decylalkohol, Caprinalkohol, 1-Hydroxydecan, Nonylcarbinol

A17288.0F A17288.AP
A17288.0FEA 81.3 EUR
A17288.0F A17288.AP
1-Decanol ≥98%
1-Decanol

Purity: 98+% CAS No.: 112-30-1 Molecular Formula: C10H22O Formula Weight: 158.29 Flash Point: 82°(179°F) Physical Form: Liquid Density: 0.828 Refractive Index: 1.4370 Appearance: Colourless Odour: Sweet Melting Point: 6° Boiling Point: 230-232° Solubility: Soluble in alcohol, ether. Insoluble in water Beilstein Reference: 1735221 Merck Reference: 14,2855 UN No.: 3082 MDL No.: MFCD00004747

Formel: H₃C(CH₂)₈CH₂OH
Molecular Weight: 158,28 g/mol
Siedepunkt: 230…232 °C
Schmelzpunkt: 6 °C
Dichte: 0,828
Flammpunkt: 82 °C (179 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00004747
CAS-Nummer: 112-30-1
EINECS: 203-956-9
UN: 3082
ADR: 9,III
Merck Index: 14,2855

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR