3-Pentanol ≥98%
Lieferant: Thermo Scientific
|
Achtung
|
L06065.14
L06065.22
L06065.36
L06065.14EA
28.5
EUR
L06065.14
L06065.22
L06065.36
3-Pentanol ≥98%
3-Pentanol
Purity: 98+%
CAS No.: 584-02-1
Molecular Formula: C5H12O
Formula Weight: 88.15
Flash Point: 33°(91°F)
Refractive Index: 1.4100
Odour: Characteristic
Melting Point: -8°
Boiling Point: 114-115°
Beilstein Reference: 1730964
Merck Reference: 14,7120
UN No.: 1105
MDL No.: MFCD00004574
Formel:
C₅H₁₂O Molecular Weight: 88,15 g/mol Siedepunkt: 114…115 °C Schmelzpunkt: –8 °C Dichte: 0,817 Flammpunkt: 33 °C (91 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00004574 CAS-Nummer: 584-02-1 EINECS: 209-526-7 UN: 1105 ADR: 3,III Merck Index: 14,7120 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...