Methanol, LiChrosolv® LC, Supelco®
Lieferant: Merck
Gefahr
Synonyme: Hydroxymethan, Methylalkohol, Holzalkohol
MQ100
1.06018.1000 1.06018.2500 1.06018.9010 1.06018.9030 1.06018.9185 1.06018.4000 1.06018.9910
1.06018.1000EA 46.4 EUR
1.06018.1000 1.06018.2500 1.06018.9010 1.06018.9030 1.06018.9185 1.06018.4000 1.06018.9910
Methanol, LiChrosolv® LC, Supelco®
Methanol
With their high degree of UV transmittance, low particle count, low acidity and alkalinity and low evaporation residue level, LiChrosolv® solvents are ideal for reproducible separations. Since separations are normally carried out under gradient conditions in analytical HPLC, we offer solvents in "gradient grade" as well as "isocratic grade". This enables to minimize the gradient effect of the solvent involved.
Formel: CH₃OH
Molecular Weight: 32.04 g/mol
Siedepunkt: 64.6
Schmelzpunkt: –98 °C
Dichte: 0.7918 g/cm³ (20 °C)
Flammpunkt: 11 °C (geschlossener Tiegel)
Lagertemperatur: Raumtemperatur
MDL: MFCD00004595
CAS-Nummer: 67-56-1
EINECS: 200-659-6
UN: 1230
ADR: 3 (6.1) II
REACH: 01-2119433307-44
Merck Index: 13,05984
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5ea983428c5015ad8417c1e1c260cce3a4d1de3c&vid=9bc2f21fdce2dcabce477372374ff56b8fb3a0f1&prid=web&clid=SAPDAM