1,5-Pentandiol ≥97%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
Pentamethylenglykol, Pentamethylenglycol, 1,5-Dihydroxypentan
A13998.0E
A13998.22
A13998.36
A13998.0EEA
183
EUR
A13998.0E
A13998.22
A13998.36
1,5-Pentandiol ≥97%
1,5-Pentandiol
Purity: 97%
CAS No.: 111-29-5
Molecular Formula: C5H12O2
Formula Weight: 104.15
Flash Point: 129°(264°F)
Refractive Index: 1.4500
Melting Point: -16°
Boiling Point: 241-242°
Beilstein Reference: 1560130
Merck Reference: 14,7117
MDL No.: MFCD00002978
Formel:
C₅H₁₂O₂ Molecular Weight: 104,15 g/mol Siedepunkt: 241…242 °C Schmelzpunkt: –16 °C Dichte: 0,992 Flammpunkt: 129 °C (264 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00002978 CAS-Nummer: 111-29-5 EINECS: 203-854-4 Merck Index: 14,7117 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...