1-Butanthiol ≥98%
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
n-Butylmercaptan, 1-Mercaptobutan, NBM, n-Butanthiol
43421.AE
43421.AP
43421.AC
43421.AEEA
48.2
EUR
43421.AE
43421.AP
43421.AC
1-Butanthiol ≥98%
1-Butanthiol
Purity: 98%
CAS No.: 109-79-5
Molecular Formula: C4H10S
Formula Weight: 90.19
Flash Point: 2°(36°F)
Physical Form: Liquid
Density: 0.842
Refractive Index: 1.4425
Sensitivity: Air Sensitive
Appearance: Colourless
Odour: Stench
Melting Point: -116°
Boiling Point: 98°
Beilstein Reference: 1730908
Merck Reference: 14,1577
UN No.: 2347
MDL No.: MFCD00004905
Formel:
C₄H₁₀S Molecular Weight: 90,19 g/mol Siedepunkt: 98 °C Schmelzpunkt: –116 °C Dichte: 0,842 Flammpunkt: 2 °C (36 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00004905 CAS-Nummer: 109-79-5 EINECS: 203-705-3 UN: 2347 ADR: 3,II Merck Index: 14,1577 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...