1-Decanthiol 96%

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: n-Decylmercaptan, 1-Mercaptodecan, n-Decanthiol

A15913.AC A15913.AE A15913.AP
A15913.ACEA 33.4 EUR
A15913.AC A15913.AE A15913.AP
1-Decanthiol 96%
1-Decanthiol

Purity: 96% CAS No.: 143-10-2 Molecular Formula: C10H22S Formula Weight: 174.35 Flash Point: 98°(208°F) Refractive Index: 1.4565 Odour: Unpleasant Melting Point: -26° Boiling Point: 237° Beilstein Reference: 1735223 MDL No.: MFCD00004884

Formel: C₁₀H₂₂S
Molecular Weight: 174.35 g/mol
Siedepunkt: 237 °C
Schmelzpunkt: –26 °C
Dichte: 0.843
Flammpunkt: 98 °C (208 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00004884
CAS-Nummer: 143-10-2
EINECS: 205-584-2
UN: 3334
ADR: 9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR