Ethanthiol ≥97%
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
1-Mercaptoethan, Ethylmercaptan
22585.AE
22585.AP
22585.AEEA
36.3
EUR
22585.AE
22585.AP
Ethanthiol ≥97%
Ethanthiol
Purity: 97%
CAS No.: 75-08-1
Molecular Formula: C2H6S
Formula Weight: 62.13
Flash Point: -48°(-55°F)
Physical Form: Liquid
Density: 0.84
Refractive Index: 1.4310
Appearance: Colourless
Odour: Unpleasant
Melting Point: -148°
Boiling Point: 35°
Solubility: Soluble in alcohol, ether, water
Beilstein Reference: 773638
Merck Reference: 14,3726
UN No.: 2363
MDL No.: MFCD00004887
Formel:
C₂H₆S Molecular Weight: 62,14 g/mol Siedepunkt: 35 °C Schmelzpunkt: –148 °C Dichte: 0,84 Flammpunkt: –48 °C (–55 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00004887 CAS-Nummer: 75-08-1 EINECS: 200-837-3 UN: 2363 ADR: 3,I Merck Index: 14,3726 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...