2-Propanthiol ≥98%

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: Isopropylmercaptan, 2-Mercaptopropan, Isopropanthiol

unter Argon abgefüllt

43422.AE 43422.AC 43422.AP
43422.AEEA 36.4 EUR
43422.AE 43422.AC 43422.AP
2-Propanthiol ≥98%
2-Propanthiol

Purity: 98% CAS No.: 75-33-2 Molecular Formula: C3H8S Formula Weight: 76.16 Flash Point: -34° (-29°F) Physical Form: Liquid, Packaged under argon Density: 0.82 Refractive Index: 1.4255 Sensitivity: Air Sensitive Appearance: Colourless Odour: Stench Melting Point: -131° Boiling Point: 57-60° Beilstein Reference: 605260 UN No.: 2402 MDL No.: MFCD00004863

Formel: C₃H₈S
Siedepunkt: 57…60 °C
Schmelzpunkt: –131 °C
Dichte: 0,82
Flammpunkt: –34° (–29 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00004863
CAS-Nummer: 75-33-2
EINECS: 200-861-4
UN: 2402
ADR: 3,II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR