Natriumdecylsulfat ≥99%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
A16299.03
A16299.06
A16299.14
A16299.03EA
40.9
EUR
A16299.03
A16299.06
A16299.14
Natriumdecylsulfat ≥99%
Natriumdecylsulfat
Purity: 99%
CAS No.: 142-87-0
Molecular Formula: CH3(CH2)9OSO3Na
Formula Weight: 260.33
Beilstein Reference: 3576860
MDL No.: MFCD00041881
Formel:
C₁₀H₂₁NaO₄S Schmelzpunkt: 191,85 °C Dichte: 1,207 g/cm³ (20 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00041881 CAS-Nummer: 142-87-0 EINECS: 205-568-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...