n-Heneicosan ≥99%

Lieferant: Thermo Fisher Scientific

A18198.03 A18198.06 A18198.14
A18198.03EA 30.2 EUR
A18198.03 A18198.06 A18198.14
n-Heneicosan ≥99%
n-Heneicosan

Purity: 99% CAS No.: 629-94-7 Molecular Formula: CH3(CH2)19CH3 Formula Weight: 296.58 Flash Point: 110°(230°F) Melting Point: 39-43° Boiling Point: 100°/2mm Beilstein Reference: 1748500 MDL No.: MFCD00009346

Formel: H₃C(CH₂)₁₉CH₃
Molecular Weight: 296.58 g/mol
Siedepunkt: 100 °C (2 mmHg)
Schmelzpunkt: 39…43 °C
Dichte: 0.792
Flammpunkt: >110 °C (230 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00009346
CAS-Nummer: 629-94-7
EINECS: 211-118-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR