1-Acetylimidazol 98%

Lieferant: Thermo Scientific

Synonyme: 1-Acetyl-1H-imidazol

169500250 169501000
ACRO169500250EA 39.3 EUR
ACRO169500250 ACRO169501000
1-Acetylimidazol 98%
1-Acetylimidazol
Formel: C₅H₆N₂O
Molecular Weight: 110,12 g/mol
Schmelzpunkt: 93…96 °C
MDL: MFCD00005287
CAS-Nummer: 2466-76-4
EINECS: 219-577-7

Bestellen

Spezifikation Testergebnisse

Appearance White to beige to tan Adhering crystalline powder or crystals
Infrared spectrum Conforms
Titration with HClO4 ≥97.5 %
NMR (1H-NMR) Consistent with structure

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR