Cobalt(II)sulfat Hydrat (≥20,8% Co)
Lieferant: Thermo Fisher Scientific
Gefahr
Synonyme: Cobaltsulfat Hydrat
11340.30 11340.A1
11340.30EA 93.7 EUR
11340.30 11340.A1
Cobalt(II)sulfat Hydrat (≥20,8% Co)
Cobalt(II)sulfat Hydrat
CAS No.: 60459-08-7 Molecular Formula: CoO4S.xH2O Formula Weight: 154.99(anhy) Physical Form: Crystalline Appearance: Red-orange Solubility: Soluble in water. Slightly soluble in methanol and ethanol MDL No.: MFCD00149657
Ceramics, pigments, glazes, plating baths, catalyst, storage batteries, paint/ink drier
Formel: CoO₄S·H₂O
Molecular Weight: 173.01 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00149657
CAS-Nummer: 60459-08-7
EINECS: 233-334-2
UN: 3077
ADR: 9 III
Merck Index: 12,02510