Ammoniumcobalt(II)sulfat Hexahydrat ≥99%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
Cobalt(II)ammoniumsulfat Hexahydrat
43902.22
43902.36
43902.22EA
140
EUR
43902.22
43902.36
Ammoniumcobalt(II)sulfat Hexahydrat ≥99%
Ammoniumcobalt(II)sulfat Hexahydrat
Purity: 99+%
CAS No.: 13586-38-4
Molecular Formula: H8CoN2O8S2.6H2O
Formula Weight: 395.22 (287.14anhy)
Physical Form: Crystalline
Density: 1.902
Appearance: Pink to red
Odour: Odourless
Melting Point: ca 120° dec.
Solubility: Soluble in water, Insoluble in alcohol
UN No.: 3077
MDL No.: MFCD00151049
As spectroscopic standard, cobalt plating, catalysis, ceramics
Formel:
H₈CoN₂O₈S₂·6H₂O Molecular Weight: 395,23 g/mol Schmelzpunkt: ∼ 120 °C (Zers.) Dichte: 1,902 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00151049 CAS-Nummer: 13586-38-4 EINECS: 237-043-1 ADR: 9,III |