trans,trans-1,4-Diphenyl-1,3-butadien ≥98%
Lieferant: Thermo Fisher Scientific
Synonyme: trans,trans-DPB, trans,trans-Bistyryl, trans,trans-PPB
A17531.06 A17531.14 A17531.22
A17531.06EA 25.6 EUR
A17531.06 A17531.14 A17531.22
trans,trans-1,4-Diphenyl-1,3-butadien ≥98%
trans,trans-1,4-Diphenyl-1,3-butadien
Purity: 98+% CAS No.: 538-81-8 Molecular Formula: C16H14 Formula Weight: 206.29 Odour: Odourless Melting Point: 151-154° Boiling Point: 350°/720mm Beilstein Reference: 1905937 Merck Reference: 14,3320 MDL No.: MFCD00004791
Formel: C₁₆H₁₄
Molecular Weight: 206.29 g/mol
Siedepunkt: 350 °C (720 mmHg)
Schmelzpunkt: 151…154 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00004791
CAS-Nummer: 538-81-8
EINECS: 212-952-6