4'-Brom-3'-nitroacetophenon ≥99%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
1-(4-Brom-3-nitrophenyl)ethan-1-on
B24469.06
B24469.14
B24469.06EA
61.4
EUR
B24469.06
B24469.14
4'-Brom-3'-nitroacetophenon ≥99%
4'-Brom-3'-nitroacetophenon
Purity: 99%
CAS No.: 18640-58-9
Molecular Formula: C8H6BrNO3
Formula Weight: 244.05
Melting Point: 117-120°
Beilstein Reference: 2050098
Formel:
C₈H₆BrNO₃ Molecular Weight: 244,04 g/mol Schmelzpunkt: 117…120 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00016985 CAS-Nummer: 18640-58-9 EINECS: 242-469-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...