Propargylamin 98%
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
3-Aminopropin
H53495.06
H53495.14
H53495.06EA
65.5
EUR
H53495.06
H53495.14
Propargylamin 98%
Propargylamin
Purity: 98%
CAS No.: 2450-71-7
Molecular Formula: C3H5N
Formula Weight: 55.08
Storage Temperature: 2°C to 8°C
Flash Point: 6°(43°F)
Refractive Index: 1.449
Boiling Point: 83°
Beilstein Reference: 773681
UN No.: 3286
MDL No.: MFCD00008198
Formel:
C₃H₅N Molecular Weight: 55,08 g/mol Siedepunkt: 83 °C Dichte: 0,86 Flammpunkt: 6 °C (43 °F) |
MDL:
MFCD00008198 CAS-Nummer: 2450-71-7 EINECS: 219-513-8 UN: 3286 ADR: 3,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...