1-Butylimidazol ≥99%

Lieferant: Thermo Scientific
Gefahr

Synonyme: N-Butyl-1H-imidazol, N-Butylimidazol, 1-Butyl-1H-imidazol

L07793.14 L07793.22 AB174528.0025
L07793.14EA 40.8 EUR
L07793.14 L07793.22 ABCRAB174528.0025
1-Butylimidazol ≥99%
1-Butylimidazole

Purity: 99% CAS No.: 4316-42-1 Molecular Formula: C7H12N2 Formula Weight: 124.19 Flash Point: 145°(293°F) Refractive Index: 1.4800 Odour: Amine-like Boiling Point: 257° Beilstein Reference: 109499 UN No.: 2690 MDL No.: MFCD00042753

Formel: C₇H₁₂N₂
Molecular Weight: 124,19 g/mol
Siedepunkt: 257 °C
Dichte: 0,948
Flammpunkt: 145 °C (293 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00042753
CAS-Nummer: 4316-42-1
EINECS: 224-335-9
UN: 2690
ADR: 6.1,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR