Propiolaldehyddiethylacetal ≥97%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
Propinaldiethylacetal, 3,3-Diethoxypropin, Propioaldehyddiethylacetal, Propargylaldehyddiethylacetal
L10767.06
L10767.14
L10767.03
L10767.06EA
146
EUR
L10767.06
L10767.14
L10767.03
Propiolaldehyddiethylacetal ≥97%
Propiolaldehyddiethylacetal
Purity: 97%
CAS No.: 10160-87-9
Molecular Formula: HC≡CCH(OCH2CH3)2
Formula Weight: 128.17
Storage Temperature: 2°C to 8°C
Flash Point: 32°(89°F)
Refractive Index: 1.4120
Boiling Point: 138-139°
Beilstein Reference: 1701566
UN No.: 1993
MDL No.: MFCD00009237
Formel:
C₇H₁₂O₂ Molecular Weight: 128,17 g/mol Siedepunkt: 138…139 °C Dichte: 0,894 Flammpunkt: 32 °C (89 °F) |
MDL:
MFCD00009237 CAS-Nummer: 10160-87-9 EINECS: 233-430-4 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...