3'-Chlor-4'-hydroxyacetophenon

Lieferant: BIONET RESEARCH

Synonyme: 1-(3-Chlor-4-hydroxyphenyl)ethan-1-on, 1-(3-Chlor-4-hydroxyphenyl)ethanon

5Y-0939.10 5Y-0939.500 5Y-0939.1 5Y-0939.5
BIOT5Y-0939.10EA 0 EUR
BIOT5Y-0939.10 BIOT5Y-0939.500 BIOT5Y-0939.1 BIOT5Y-0939.5
3'-Chlor-4'-hydroxyacetophenon
3'-Chlor-4'-hydroxyacetophenon
Formel: C₈H₇ClO₂
Molecular Weight: 170.6
MDL: MFCD00663621
CAS-Nummer: 2892-29-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR