4-Brom-2-methyl-6-nitroanilin

Lieferant: BIONET RESEARCH

Synonyme: 2-Amino-5-brom-3-nitrotoluol, 4-Brom-6-nitro-o-toluidin

9T-0039.500 9T-0039.1 9T-0039.5 9T-0039.10
BIOT9T-0039.500EA 0 EUR
BIOT9T-0039.500 BIOT9T-0039.1 BIOT9T-0039.5 BIOT9T-0039.10
4-Brom-2-methyl-6-nitroanilin
4-Brom-2-methyl-6-nitroanilin
Formel: C₇H₇BrN₂O₂
Molecular Weight: 231.05 g/mol
Schmelzpunkt: 155
MDL: MFCD00052919
CAS-Nummer: 77811-44-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR