4-Brom-2-methyl-6-nitroanilin

Lieferant: Thermo Scientific

Synonyme: 2-Amino-5-brom-3-nitrotoluol, 4-Brom-6-nitro-o-toluidin

BTB07466.1 BTB07466.10 BTB07466.50
MAYBBTB07466.1EA 0 EUR
MAYBBTB07466.1 MAYBBTB07466.10 MAYBBTB07466.50
4-Brom-2-methyl-6-nitroanilin
4-Brom-2-methyl-6-nitroanilin
Formel: C₇H₇BrN₂O₂
Molecular Weight: 231,05 g/mol
Schmelzpunkt: 155 °C
MDL: MFCD00052919
CAS-Nummer: 77811-44-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR