2-Brom-4-methyl-6-nitroanilin

Lieferant: Thermo Scientific

Synonyme: 2-Brom-6-nitro-p-toluidin, 4-Amino-3-brom-5-nitrotoluol

BTB10711.250 BTB10711.1
MAYBBTB10711.250EA 0 EUR
MAYBBTB10711.250 MAYBBTB10711.1
2-Brom-4-methyl-6-nitroanilin
2-Brom-4-methyl-6-nitroanilin
Formel: C₇H₇BrN₂O₂
MDL: MFCD00209452
CAS-Nummer: 827-24-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR