1-Decyl-2-methylimidazol

Lieferant: CHEMPUR

BTB14163.250 BTB14163.1
CHMPBTB14163.250EA 0 EUR
CHMPBTB14163.250 MAYBBTB14163.1
1-Decyl-2-methylimidazol
1-Decyl-2-methylimidazole
Formel: C₁₄H₂₆N₂
CAS-Nummer: 42032-30-4
EINECS: 255-633-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR